COMH0423867A Combi-Blocks, Inc.

3-(4-Chlorophenyl)-1-phenyl-1h-pyrazole-5-carboxylic acid

  • MDL number MFCD05150613
  • Cas Number : 618102-33-3
  • Molecular Weight : 298.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5635

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5635
Availability details for
Quantity
  • Availability
  • Add to Cart