COMH042386D0 Combi-Blocks, Inc.

Ethyl 4-(4-bromophenyl)-2-[(cyanoacetyl)amino]thiophene-3-carboxylate

  • MDL number MFCD03392237
  • Cas Number : 546064-73-7
  • Molecular Weight : 393.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5723

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5723
Availability details for
Quantity
  • Availability
  • Add to Cart