COMH042386D9 Combi-Blocks, Inc.

3-[Ethyl(methyl)amino]propanenitrile

  • MDL number MFCD11129793
  • Cas Number : 55619-09-5
  • Molecular Weight : 112.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5732

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5732
Availability details for
Quantity
  • Availability
  • Add to Cart