COMH042386E0 Combi-Blocks, Inc.

N-(4-Methoxybenzyl)-n-[2-(5-methoxy-1h-indol-3-yl)ethyl]amine

  • MDL number MFCD01475061
  • Cas Number : 155639-31-9
  • Molecular Weight : 310.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5739

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5739
Availability details for
Quantity
  • Availability
  • Add to Cart