COMH042386FF Combi-Blocks, Inc.

2,2-Diallylazetidine

  • MDL number MFCD08445724
  • Cas Number : 889939-55-3
  • Molecular Weight : 137.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5770

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5770
Availability details for
Quantity
  • Availability
  • Add to Cart