COMH0423872B Combi-Blocks, Inc.

N-[Amino(imino)methyl]-4-bromobenzenesulfonamide

  • MDL number MFCD01882526
  • Cas Number : 36397-58-7
  • Molecular Weight : 278.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5814

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5814
Availability details for
Quantity
  • Availability
  • Add to Cart