COMH0423874A Combi-Blocks, Inc.

3-(1H-Imidazol-1-ylmethyl)-4-isopropoxybenzaldehyde

  • MDL number MFCD03180287
  • Cas Number : 947012-62-6
  • Molecular Weight : 244.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5845

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5845
Availability details for
Quantity
  • Availability
  • Add to Cart