COMH0423876D Combi-Blocks, Inc.

[(6-Fluoro-1,3-benzoxazol-2-yl)methyl]amine

  • MDL number MFCD10696598
  • Cas Number : 944907-47-5
  • Molecular Weight : 166.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5880

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5880
Availability details for
Quantity
  • Availability
  • Add to Cart