COMH0423877F Combi-Blocks, Inc.

8-Ethyl-4,4,6-trimethyl-6-phenyl-5,6-dihydro-4h-pyrrolo[3,2,1-ij]quinoline-1,2-dione

  • MDL number MFCD05843903
  • Cas Number : 835891-49-1
  • Molecular Weight : 333.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5898

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5898
Availability details for
Quantity
  • Availability
  • Add to Cart