COMH04238783 Combi-Blocks, Inc.

4-Methyl-5-(5,6,7,8-tetrahydro-4h-cyclohepta[b]thien-3-yl)-4h-1,2,4-triazole-3-thiol

  • MDL number MFCD09971977
  • Cas Number : 956576-83-3
  • Molecular Weight : 265.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5902

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5902
Availability details for
Quantity
  • Availability
  • Add to Cart