COMH04238799 Combi-Blocks, Inc.

6-(Morpholin-4-ylcarbonyl)-2,3,4,9-tetrahydro-1h-carbazole

  • MDL number MFCD09371754
  • Cas Number : 931666-01-2
  • Molecular Weight : 284.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5925

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5925
Availability details for
Quantity
  • Availability
  • Add to Cart