COMH042387B9 Combi-Blocks, Inc.

Sodium 5-tert-butyl-4-methyl-1,3-thiazole-2-carboxylate

  • MDL number MFCD18064651
  • Cas Number : 937663-35-9
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5957

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5957
Availability details for
Quantity
  • Availability
  • Add to Cart