COMH042387D4 Combi-Blocks, Inc.

1-Piperidineacetic acid, alpha,3,5-trimethyl-, hydrochloride

  • MDL number MFCD28505087
  • Cas Number : 1922694-67-4
  • Molecular Weight : 221.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5986

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5986
Availability details for
Quantity
  • Availability
  • Add to Cart