COMH042387E7 Combi-Blocks, Inc.

2-([(5-Chloro-2-methoxyphenyl)amino]methyl)-1h-isoindole-1,3(2h)-dione

  • MDL number MFCD00106892
  • Cas Number : 859299-43-7
  • Molecular Weight : 316.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6006

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6006
Availability details for
Quantity
  • Availability
  • Add to Cart