COMH042387E8 Combi-Blocks, Inc.

Ethyl 4-oxo-1,4-dihydropyrimido[1,2-a]benzimidazole-3-carboxylate

  • MDL number MFCD02615366
  • Cas Number : 40519-93-5
  • Molecular Weight : 257.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6007

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6007
Availability details for
Quantity
  • Availability
  • Add to Cart