COMH042387EE Combi-Blocks, Inc.

N-Benzyl-1-(1-ethyl-1h-imidazol-2-yl)methanamine dihydrochloride

  • MDL number MFCD18071324
  • Cas Number : 886505-56-2
  • Molecular Weight : 288.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6013

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6013
Availability details for
Quantity
  • Availability
  • Add to Cart