COMH0423880D Combi-Blocks, Inc.

2-[(3-Bromobenzoyl)amino]-4-chlorobenzoic acid

  • MDL number MFCD01811223
  • Cas Number : 827325-60-0
  • Molecular Weight : 354.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6046

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6046
Availability details for
Quantity
  • Availability
  • Add to Cart