COMH04238831 Combi-Blocks, Inc.

4-[4-(Methylthio)phenyl]-1,4-dihydro[1,3,5]triazino[1,2-a]benzimidazol-2-amine

  • MDL number MFCD06603439
  • Cas Number : 452285-32-4
  • Molecular Weight : 309.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6082

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6082
Availability details for
Quantity
  • Availability
  • Add to Cart