COMH04238843 Combi-Blocks, Inc.

Isopropyl 2-amino-4-(4-isopropylphenyl)thiophene-3-carboxylate

  • MDL number MFCD01923355
  • Cas Number : 351157-20-5
  • Molecular Weight : 303.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6100

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6100
Availability details for
Quantity
  • Availability
  • Add to Cart