COMH04238859 Combi-Blocks, Inc.

2-Bromo-n-[4-(trifluoromethyl)benzyl]acetamide

  • MDL number MFCD18856495
  • Cas Number : 1007229-11-9
  • Molecular Weight : 296.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6122

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6122
Availability details for
Quantity
  • Availability
  • Add to Cart