COMH0423885B Combi-Blocks, Inc.

[(4-Methyl-1,2,3-thiadiazol-5-yl)methyl]amine hydrochloride

  • MDL number MFCD22207116
  • Cas Number : 790641-72-4
  • Molecular Weight : 165.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6124

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6124
Availability details for
Quantity
  • Availability
  • Add to Cart