COMH04238876 Combi-Blocks, Inc.

1-Chloronaphtho[2,1-b]thiophene-2-carbonyl chloride

  • MDL number MFCD08361745
  • Cas Number : 85992-25-2
  • Molecular Weight : 281.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6151

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6151
Availability details for
Quantity
  • Availability
  • Add to Cart