COMH0423888F Combi-Blocks, Inc.

3-Bromo-1-(methoxymethyl)-n,n-dimethyl-1h-1,2,4-triazol-5-amine

  • MDL number MFCD30099284
  • Cas Number : 2090280-13-8
  • Molecular Weight : 235.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6176

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6176
Availability details for
Quantity
  • Availability
  • Add to Cart