COMH042388B6 Combi-Blocks, Inc.

3-(1,3-Benzodioxol-5-yloxy)-2-hydroxypropanoic acid

  • MDL number MFCD08282886
  • Cas Number : 924831-74-3
  • Molecular Weight : 226.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6216

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6216
Availability details for
Quantity
  • Availability
  • Add to Cart