COMH042388C3 Combi-Blocks, Inc.

1-(4-Bromo-2-fluorophenyl)-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD09811892
  • Cas Number : 1016697-56-5
  • Molecular Weight : 302.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6229

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6229
Availability details for
Quantity
  • Availability
  • Add to Cart