COMH042388F5 Combi-Blocks, Inc.

2-(2-Bromobenzoyl)-n-methylhydrazinecarbothioamide

  • MDL number MFCD03942244
  • Cas Number : 58064-69-0
  • Molecular Weight : 288.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6280

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6280
Availability details for
Quantity
  • Availability
  • Add to Cart