COMH042388F9 Combi-Blocks, Inc.

2-Amino-n-cyclohexyl-5,6,7,8-tetrahydro-4h-cyclohepta[b]thiophene-3-carboxamide

  • MDL number MFCD03422676
  • Cas Number : 438236-24-9
  • Molecular Weight : 292.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6284

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6284
Availability details for
Quantity
  • Availability
  • Add to Cart