COMH0423891C Combi-Blocks, Inc.

1-(5-Isopropyl-1,3,4-thiadiazol-2-yl)-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD30723514
  • Cas Number : 2173108-88-6
  • Molecular Weight : 255.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6320

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6320
Availability details for
Quantity
  • Availability
  • Add to Cart