COMH04238925 Combi-Blocks, Inc.

Methyl [1-(2-furoyl)piperidin-4-yl]acetate

  • MDL number MFCD13248772
  • Cas Number : 952934-82-6
  • Molecular Weight : 251.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6329

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6329
Availability details for
Quantity
  • Availability
  • Add to Cart