COMH0423893D Combi-Blocks, Inc.

(2Z)-2-(2-Chloro-6-fluorobenzylidene)-6-hydroxy-1-benzofuran-3(2h)-one

  • MDL number MFCD04181737
  • Cas Number : 637753-85-6
  • Molecular Weight : 290.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6353

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6353
Availability details for
Quantity
  • Availability
  • Add to Cart