COMH04238941 Combi-Blocks, Inc.

3-(1-[2-(3-Fluorophenyl)ethyl]-2,5-dioxoimidazolidin-4-yl)propanoic acid

  • MDL number MFCD29051089
  • Cas Number : 1955494-04-8
  • Molecular Weight : 294.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6357

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6357
Availability details for
Quantity
  • Availability
  • Add to Cart