COMH0423894F Combi-Blocks, Inc.

4-Methyl-3-[(2-thienylacetyl)amino]benzoic acid

  • MDL number MFCD01434376
  • Cas Number : 903746-53-2
  • Molecular Weight : 275.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6371

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6371
Availability details for
Quantity
  • Availability
  • Add to Cart