COMH04238993 Combi-Blocks, Inc.

5-[1-(Ethylsulfonyl)piperidin-3-yl]-4-propyl-4h-1,2,4-triazole-3-thiol

  • MDL number MFCD08569918
  • Cas Number : 932918-82-6
  • Molecular Weight : 318.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6439

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6439
Availability details for
Quantity
  • Availability
  • Add to Cart