COMH042389B5 Combi-Blocks, Inc.

3-([1,2,4]Triazolo[4,3-a]pyridin-3-yl)propan-1-amine hydrochloride

  • MDL number MFCD16040048
  • Cas Number : 610276-38-5
  • Molecular Weight : 212.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6476

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6476
Availability details for
Quantity
  • Availability
  • Add to Cart