COMH042389C0 Combi-Blocks, Inc.

4-[[2-(Azepan-1-ylsulfonyl)ethyl](pyridin-4-ylmethyl)amino]-4-oxobutanoic acid

  • MDL number MFCD08746794
  • Cas Number : 919748-82-6
  • Molecular Weight : 397.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6487

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6487
Availability details for
Quantity
  • Availability
  • Add to Cart