COMH042389E2 Combi-Blocks, Inc.

N-(3-Chlorophenyl)-n'-[2-hydroxy-1,1-bis(hydroxymethyl)ethyl]urea

  • MDL number MFCD00589818
  • Cas Number : 448189-13-7
  • Molecular Weight : 274.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6521

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6521
Availability details for
Quantity
  • Availability
  • Add to Cart