COMH042389EB Combi-Blocks, Inc.

2-(3-Pyrimidin-4-ylpropyl)azetidine-2-carboxamide bis(trifluoroacetate)

  • MDL number MFCD29035024
  • Cas Number : 2108655-91-8
  • Molecular Weight : 448.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6530

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6530
Availability details for
Quantity
  • Availability
  • Add to Cart