COMH042389F4 Combi-Blocks, Inc.

4-Propoxypyrimidine-2-thiol

  • MDL number MFCD00071712
  • Cas Number : 438219-92-2
  • Molecular Weight : 170.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6539

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6539
Availability details for
Quantity
  • Availability
  • Add to Cart