COMH04238A31 Combi-Blocks, Inc.

2-Chloro-n-(3,4,5-trimethoxyphenyl)acetamide

  • MDL number MFCD00524910
  • Cas Number : 39901-45-6
  • Molecular Weight : 259.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6601

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6601
Availability details for
Quantity
  • Availability
  • Add to Cart