COMH04238A42 Combi-Blocks, Inc.

Ethyl 3-amino-3-(4-isopropoxy-3-methoxyphenyl)propanoate hydrochloride

  • MDL number MFCD12027412
  • Cas Number : 945451-04-7
  • Molecular Weight : 317.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6618

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6618
Availability details for
Quantity
  • Availability
  • Add to Cart