COMH04238A5A Combi-Blocks, Inc.

3-(2-Furyl)-1h-1,2,4-triazole

  • MDL number MFCD18784752
  • Cas Number : 23195-65-5
  • Molecular Weight : 135.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6642

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6642
Availability details for
Quantity
  • Availability
  • Add to Cart