COMH04238A6C Combi-Blocks, Inc.

N-[1-(Aminomethyl)-4-methylcyclohexyl]-n,n-dimethylamine hydrochloride

  • MDL number MFCD22207125
  • Cas Number : 919013-75-5
  • Molecular Weight : 206.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6661

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6661
Availability details for
Quantity
  • Availability
  • Add to Cart