COMH04238A9B Combi-Blocks, Inc.

4-(4-Nitro-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl)benzoic acid

  • MDL number MFCD00391880
  • Cas Number : 71494-65-0
  • Molecular Weight : 312.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6708

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6708
Availability details for
Quantity
  • Availability
  • Add to Cart