COMH04238AA6 Combi-Blocks, Inc.

N-(2-Bromophenyl)-3-chloropropanamide

  • MDL number MFCD03386570
  • Cas Number : 545364-03-2
  • Molecular Weight : 262.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6720

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6720
Availability details for
Quantity
  • Availability
  • Add to Cart