COMH04238AAA Combi-Blocks, Inc.

2-Morpholin-4-yl-2-pyridin-3-ylacetamide

  • MDL number MFCD03761249
  • Cas Number : 719279-71-7
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6724

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6724
Availability details for
Quantity
  • Availability
  • Add to Cart