COMH04238AB9 Combi-Blocks, Inc.

N-(2-[(5-Nitroquinolin-8-yl)oxy]ethyl)acetamide

  • MDL number MFCD05854566
  • Cas Number : 696607-18-8
  • Molecular Weight : 275.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6739

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6739
Availability details for
Quantity
  • Availability
  • Add to Cart