COMH04238AD2 Combi-Blocks, Inc.

1-Phenyladamantane

  • MDL number MFCD00154936
  • Cas Number : 780-68-7
  • Molecular Weight : 212.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6764

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6764
Availability details for
Quantity
  • Availability
  • Add to Cart