COMH04238B33 Combi-Blocks, Inc.

8-Chloro-2-(5-methylthien-2-yl)quinoline-4-carboxylic acid

  • MDL number MFCD03422167
  • Cas Number : 725221-34-1
  • Molecular Weight : 303.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6862

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6862
Availability details for
Quantity
  • Availability
  • Add to Cart