COMH04238B4B Combi-Blocks, Inc.

Diethyl 2,2'-tricyclo[3.3.1.1(3,7)]decane-1,3-diyldiacetate

  • MDL number MFCD09743482
  • Cas Number : 81657-07-0
  • Molecular Weight : 308.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6887

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6887
Availability details for
Quantity
  • Availability
  • Add to Cart