COMH04238B4D Combi-Blocks, Inc.

2-Methyl-5-nitro-3-phenyl-1h-indole

  • MDL number MFCD00563936
  • Cas Number : 41018-87-5
  • Molecular Weight : 252.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6889

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6889
Availability details for
Quantity
  • Availability
  • Add to Cart